missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%, Thermo Scientific Chemicals

Catalog Number p-3778737
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
375-01-9
C4H3F7O
200.06
MFCD00004674
WXJFKAZDSQLPBX-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
9776
2,2,3,3,4,4,4-heptafluorobutan-1-ol
OCC(F)(F)C(F)(F)C(F)(F)F
This item is not returnable. View return policy
375-01-9
C4H3F7O
200.06
MFCD00004674
WXJFKAZDSQLPBX-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
9776
2,2,3,3,4,4,4-heptafluorobutan-1-ol
OCC(F)(F)C(F)(F)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Density 1.6000g/mL
Boiling Point 96°C to 97°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CF3CF2CF2CH2OH
Refractive Index 1.2920 to 1.294
Quantity 10 g
Beilstein 01, 1547
Specific Gravity 1.6
Solubility Information Solubility in water: slightly soluble
Formula Weight 200.06
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,3,3,4,4,4-Heptafluoro-1-butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'