Få mere at vide
2,2',3,3',4,3',6-Heptachlorobiphenyl, SPEX CertiPrep™
Artikelnummer. 16300088 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
16300088 1 mL
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 16300088 Leverandør SPEX Certiprep Leverandørnr. S2107
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
2,2',3,3',4,4',6-Heptachlorobiphenyl52663-71-51000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale 2,2',3,3',4,4',6-Heptachlorobiphenyl
Emballage 2mL amber prescored glass ampule
CAS 52663-71-5, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C12H3Cl7
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle TZMHVHLTPWKZCI-UHFFFAOYSA-N
SMIL C1=CC(=C(C(=C1C2=C(C(=C(C=C2Cl)Cl)Cl)Cl)Cl)Cl)Cl
IUPAC navn 1,2,3,5-tetrachloro-4-(2,3,4-trichlorophenyl)benzene
Opløsningsmiddel eller Matrix Methanol-P&T
Molekylvægt (g/mol) 395.306
PubChem CID 40478
Mængde 1 mL
Vis mere Vis mindre
Product intended for laboratory use only