missing translation for 'onlineSavingsMsg'
Få mere at vide

2,2,2-Trifluoroethyl perfluorobutanesulphonate, TRC

Artikelnummer. 30450193
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450193 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30450193 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7905751G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2,2,2-Trifluoroethyl perfluorobutanesulphonate
CAS 79963-95-4
Molekylær formel C6H2F12O3S
Mængde 1 g
Synonym 1,1,2,2,3,3,4,4,4-Nonafluorobutane-1-sulfonic Acid 2,2,2-trifluoroethyl Ester, 2,2,2-Trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate, 2,2,2-Trifluoroethyl Nonafluorobutanesulfonate
InChI formel InChI=1S/C6H2F12O3S/c7-2(8,9)1-21-22(19,20)6(17,18)4(12,13)3(10,11)5(14,15)16/h1H2
SMIL FC(F)(F)COS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC navn 2,2,2-trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
Molekylvægt (g/mol) 382.12
Formel vægt 381.9533
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.