Learn More
2,2,2-Trifluoroethanol, Sigma-Aldrich™
Product Code. 15879570
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879570 250 mL 250mL
15869570 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879570 Supplier Sigma Aldrich Supplier No. 8082590250
 Restricted Item
Forventet forsendelse: 19-11-2026
For synthesis
2,2,2-Trifluoroethanol (TFE) is a fluorinated compound used as a solvent in organic synthesis. Peptides and proteins are experimentally studied using TEE as a cosolvent. In TFE, hydrogen peroxide is effective for oxidizing sulfur compounds.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-89-8
Grade Research
Molecular Weight (g/mol) 100.04
Specifications
Melting Point -43.5°C
Density 1.38 g/cm3 at 25°C
Boiling Point 73.6°C/1013 hPa
MDL Number MFCD00004672
Quantity 250 mL
Synonym 2,2,2-Trifluoroethanol, 2,2,2-Trifluoroethanol