Få mere at vide
2-[2-(2-t-Boc-aminoethoxy]ethoxy]ethyl Bromide, TRC
Artikelnummer. 30345104
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30345104 500 mg
30343914 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30345104 Leverandør Toronto Research Chemicals Leverandørnr. TRCB618590500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[2-(2-t-Boc-aminoethoxy]ethoxy]ethyl Bromide
CAS 165963-71-3
Molekylær formel C11 H22 Br N O4
Mængde 500 mg
Synonym 2,2′-[Oxybis(2,1-ethanediyloxy)]bis[ethanol], 1,11-Dihydroxy-3,6,9-trioxaundecane, 2,2′-((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethanol), 2,2′-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, 2,2′-[Oxybis(2,1-ethanediyloxy)]diethanol, 2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethan-1-ol, 3,6,9-Trioxaundecane-1,11-diol, Bis[2-(2-hydroxyethoxy)ethyl] Ether, Hi-Dry, NSC 1262, TEG
InChI formel InChI=1S/C11H22BrNO4/c1-11(2,3)17-10(14)13-5-7-16-9-8-15-6-4-12/h4-9H2,1-3H3,(H,13,14)
SMIL CC(C)(C)OC(=O)NCCOCCOCCBr
IUPAC navn tert-butyl N-[2-[2-(2-bromoethoxy)ethoxy]ethyl]carbamate
Molekylvægt (g/mol) 312.2007
Formel vægt 311.0732