Få mere at vide
2-[2-(2-Butoxyethoxy)ethoxy]tetrahydropyran-d9, TRC
Artikelnummer. 30347917
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30347917 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347917 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6905875MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[2-(2-Butoxyethoxy)ethoxy]tetrahydropyran-d9
CAS 1346598-46-6
Molekylær formel C13H17D9O4
Mængde 5 mg
Synonym 2-[2-(2-butoxyethoxy)ethoxy]tetrahydro-2H-pyran-d9
InChI formel InChI=1S/C13H26O4/c1-2-3-7-14-9-10-15-11-12-17-13-6-4-5-8-16-13/h13H,2-12H2,1H3/i1D3,2D2,3D2,7D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OCCOCCOC1CCCCO1
IUPAC navn 2-[2-[2-(1,1,2,2,3,3,4,4,4-nonadeuteriobutoxy)ethoxy]ethoxy]oxane
Molekylvægt (g/mol) 255.4
Formel vægt 255.2396