Få mere at vide
2-[2-(2-Aminophenyl)vinyl]quinolin-8-ol (>90%), TRC
Artikelnummer. 30308897
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308897 100 mg
30309320 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30308897 Leverandør Toronto Research Chemicals Leverandørnr. TRCA615795100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[2-(2-Aminophenyl)vinyl]quinolin-8-ol
CAS 429651-60-5
Molekylær formel C17 H14 N2 O
Mængde 100 mg
InChI formel InChI=1S/C17H14N2O/c18-15-6-2-1-4-12(15)8-10-14-11-9-13-5-3-7-16(20)17(13)19-14/h1-11,20H,18H2
SMIL Nc1ccccc1C=Cc2ccc3cccc(O)c3n2
IUPAC navn 2-[2-(2-aminophenyl)ethenyl]quinolin-8-ol
Molekylvægt (g/mol) 262.306
Formel vægt 262.111