Få mere at vide
2,2,14,14-Tetramethyl-8-oxopentadecanedioic Acid-d4, TRC
Artikelnummer. 30453764
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30453764 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453764 Leverandør Toronto Research Chemicals Leverandørnr. TRCT48241250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2,14,14-Tetramethyl-8-oxopentadecanedioic Acid-d4
CAS 2408132-01-2
Molekylær formel C19 D4 H30 O5
Mængde 50 mg
Synonym 8-Oxo-2,2,14,14-tetramethylpentadecanedioic Acid-d4, ESP 15228-d4
InChI formel InChI=1S/C19H34O5/c1-18(2,16(21)22)13-9-5-7-11-15(20)12-8-6-10-14-19(3,4)17(23)24/h5-14H2,1-4H3,(H,21,22)(H,23,24)/i11D2,12D2
SMIL [2H]C([2H])(CCCCC(C)(C)C(=O)O)C(=O)C([2H])([2H])CCCCC(C)(C)C(=O)O
IUPAC navn 7,7,9,9-tetradeuterio-2,2,14,14-tetramethyl-8-oxopentadecanedioic acid
Molekylvægt (g/mol) 346.495
Formel vægt 346.266