missing translation for 'onlineSavingsMsg'
Learn More

[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol, Mikromol™

Product Code. 30050930
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30050930 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050930 Supplier Mikromol Supplier No. MM0168.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2'-(1H-Tetrazol-5-yl)biphenyl-4-yl)methanol
CAS 160514-13-6
Analyt- eller komponentnavne (2'-(1H-Tetrazol-5-yl)biphenyl-4-yl)methanol
Forsendelsestilstand Room Temperature
Molekylær formel C14 H12 N4 O
Certificeringer/compliance ISO 17025
Synonym Losartan Potassium Imp. B (EP), Losartan Imp. B (EP), [2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol, Losartan Potassium Impurity B, Losartan Impurity B
InChI formel InChI=1S/C14H12N4O/c19-9-10-5-7-11(8-6-10)12-3-1-2-4-13(12)14-15-17-18-16-14/h1-8,19H,9H2,(H,15,16,17,18)
SMIL OCc1ccc(cc1)c2ccccc2c3nnn[nH]3
IUPAC navn [4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methanol
Molekylvægt (g/mol) 252.27
Mængde 25 mg
Formel vægt 252.1011 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.