Learn More
Alfa Aesar™ 2(1H)-Quinolinone-3-carboxaldehyde, 97%
Brand: Alfa Aesar™ H33508.03
Additional Details : CAS Number : 91301-03-0 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
91301-03-0 | |
173.171 | |
VWHKEYXRRNSJTN-UHFFFAOYSA-N | |
589334 | |
C1=CC=C2C(=C1)C=C(C(=O)N2)C=O |
C10H7NO2 | |
MFCD00563443 | |
2-hydroxyquinoline-3-carbaldehyde, 2-oxo-1,2-dihydro-quinoline-3-carbaldehyde, 2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 3-formyl-2quinolone, 1,2-dihydro-2-oxoquinoline-3-carboxaldehyde, 1,2-dihydro-2-oxoquinoline-3-carbaldehyde, 2-hydroxy-3-quinolinecarboxaldehyde, 2-oxo-1,2-dihydro-3-quinolinecarbaldehyde, 2-oxohydroquinoline-3-carbaldehyde, 3-formylcarbostyril | |
2-oxo-1H-quinoline-3-carbaldehyde |
Specifications
2(1H)-Quinolinone-3-carboxaldehyde | |
White to Brown | |
1g | |
C10H7NO2 | |
2-hydroxyquinoline-3-carbaldehyde, 2-oxo-1,2-dihydro-quinoline-3-carbaldehyde, 2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 3-formyl-2quinolone, 1,2-dihydro-2-oxoquinoline-3-carboxaldehyde, 1,2-dihydro-2-oxoquinoline-3-carbaldehyde, 2-hydroxy-3-quinolinecarboxaldehyde, 2-oxo-1,2-dihydro-3-quinolinecarbaldehyde, 2-oxohydroquinoline-3-carbaldehyde, 3-formylcarbostyril | |
C1=CC=C2C(=C1)C=C(C(=O)N2)C=O | |
173.171 | |
173.2 | |
97% |
Air Sensitive | |
>250°C | |
91301-03-0 | |
MFCD00563443 | |
VWHKEYXRRNSJTN-UHFFFAOYSA-N | |
2-oxo-1H-quinoline-3-carbaldehyde | |
589334 | |
Solid |