Få mere at vide
2-(1H-Pyrazol-4-yl)aniline, TRC
Artikelnummer. 30337654
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337654 10 mg
30340539 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30337654 Leverandør Toronto Research Chemicals Leverandørnr. TRCB53020010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(1H-Pyrazol-4-yl)aniline
CAS 1342757-33-8
Molekylær formel C9H9N3
Mængde 10 mg
Synonym 2-(1H-pyrazol-4-yl)aniline dihydrochloride
InChI formel InChI=1S/C9H9N3/c10-9-4-2-1-3-8(9)7-5-11-12-6-7/h1-6H,10H2,(H,11,12)
SMIL Nc1ccccc1c2cn[nH]c2
IUPAC navn 2-(1H-pyrazol-4-yl)aniline
Molekylvægt (g/mol) 159.18
Formel vægt 159.0796