Få mere at vide
2-(1H-Indazol-6-ylsulfinyl)-N-methylbenzamide, TRC
Artikelnummer. 30404973
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30404973 100 mg
30407065 50 mg
30405532 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30404973 Leverandør Toronto Research Chemicals Leverandørnr. TRCI628102100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1959572-97-4
Molekylær formel C15H13N3O2S
Molekylvægt (g/mol) 299.35
Synonym Axitinib Impurity C
IUPAC navn 2-(1H-indazol-6-ylsulfinyl)-N-methylbenzamide
SMIL CNC(=O)c1ccccc1S(=O)c2ccc3cn[nH]c3c2
Tekniske data
Kemisk navn eller materiale 2-(1H-Indazol-6-ylsulfinyl)-N-methylbenzamide
Mængde 100 mg
InChI formel InChI=1S/C15H13N3O2S/c1-16-15(19)12-4-2-3-5-14(12)21(20)11-7-6-10-9-17-18-13(10)8-11/h2-9H,1H3,(H,16,19)(H,17,18)
Formel vægt 299.0728