missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperidinyl)phenol, 98%, Thermo Scientific Chemicals

Catalog Number 11331997
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
65195-20-2
C11H15NO
177.25
MFCD00051687
OXELIFPFCCGAJX-UHFFFAOYSA-N
2-piperidin-1-yl phenol, 2-piperidinophenol, 2-1-piperidino phenol, 2-1-piperidinyl phenol, 2-piperidylphenol, 2-hydroxyphenyl piperidine, 1-2-hydroxyphenyl piperidine
3262250
OC1=CC=CC=C1N1CCCCC1
This item is not returnable. View return policy
65195-20-2
C11H15NO
177.25
MFCD00051687
OXELIFPFCCGAJX-UHFFFAOYSA-N
2-piperidin-1-yl phenol, 2-piperidinophenol, 2-1-piperidino phenol, 2-1-piperidinyl phenol, 2-piperidylphenol, 2-hydroxyphenyl piperidine, 1-2-hydroxyphenyl piperidine
3262250
OC1=CC=CC=C1N1CCCCC1

2-(1-Piperidinyl)phenol is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(1-Piperidinyl)phenol is used as primary and secondary intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 73°C to 76°C
Quantity 1 g
Beilstein 151777
IUPAC Name 2-(piperidin-1-yl)phenol
Formula Weight 177.24
Percent Purity 98%
Chemical Name or Material 2-(1-Piperidinyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'