Få mere at vide
2-(1-Piperazinyl)pyrimidine-d8, TRC
Artikelnummer. 30411770
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30411770 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411770 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4813022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(1-Piperazinyl)pyrimidine-d8
CAS 1309283-31-5
Renhedsgrad noter HPLC
Molekylær formel C8H4D8N4
Mængde 2.5 mg
Synonym 1-(2-Pyrimidinyl)piperazine-d8, 2-Piperazinopyrimidine-d8, 4-(2-Pyrimidinyl)piperazine-d8, CM 56324H-d8, MJ 13653-d8, N-2-Pyrimidinylpiperazine-d8, PmP-d8, 1-PP-d8
InChI formel InChI=1S/C8H12N4/c1-2-10-8(11-3-1)12-6-4-9-5-7-12/h1-3,9H,4-7H2/i4D2,5D2,6D2,7D2
SMIL [2H]C(C([2H])([2H])NC([2H])([2H])C1([2H])[2H])([2H])N1C2=NC=CC=N2
Molekylvægt (g/mol) 172.26
Procent renhed >95