missing translation for 'onlineSavingsMsg'
Få mere at vide

2-[1-(Methylamino)ethyl]indole, TRC

Produktkod. 30465797
Change view
Klicka för att se tillgängliga alternativ
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde
30465797 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30465797 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM286150500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemisk navn eller materiale 2-[1-(Methylamino)ethyl]indole
CAS 96286-08-7
Molekylær formel C11 H14 N2
Mængde 500 mg
Synonym 2-[1-(Methylamino)ethyl] Indole
InChI formel InChI=1S/C11H14N2/c1-8(12-2)11-7-9-5-3-4-6-10(9)13-11/h3-8,12-13H,1-2H3
SMIL CNC(C)c1cc2ccccc2[nH]1
IUPAC navn 1-(1H-indol-2-yl)-N-methylethanamine
Molekylvægt (g/mol) 174.24
Formel vægt 174.1157
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.