Få mere at vide
2-(1-Amino-1-methylethyl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide-d3, TRC
Artikelnummer. 30308767
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30308767 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308767 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6161521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(1-Amino-1-methylethyl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide-d3
CAS 1189508-85-7
Molekylær formel C16H16D3FN4O3
Mængde 1 mg
Synonym 2-(1-Amino-1-methylethyl)-N-[(4-fluorophenyl)methyl]-1,6-dihydro-5-hydroxy-1-(methyl-d3)-6-oxo-4-pyrimidinecarboxamide
InChI formel InChI=1S/C16H19FN4O3/c1-16(2,18)15-20-11(12(22)14(24)21(15)3)13(23)19-8-9-4-6-10(17)7-5-9/h4-7,22H,8,18H2,1-3H3,(H,19,23)/i3D3
SMIL [2H]C([2H])([2H])N1C(=O)C(=C(N=C1C(C)(C)N)C(=NCc2ccc(F)cc2)O)O
IUPAC navn 2-(2-aminopropan-2-yl)-N-[(4-fluorophenyl)methyl]-5-hydroxy-6-oxo-1-(trideuteriomethyl)pyrimidine-4-carboxamide
Molekylvægt (g/mol) 337.36