Få mere at vide
2-[1-(4-Piperonyl)piperazinyl]benzothiazole, TRC
Artikelnummer. 30428583
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428583 10 mg
30422956 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30428583 Leverandør Toronto Research Chemicals Leverandørnr. TRCP49050310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[1-(4-Piperonyl)piperazinyl]benzothiazole
CAS 155106-73-3
Molekylær formel C19 H19 N3 O2 S
Mængde 10 mg
InChI formel InChI=1S/C19H19N3O2S/c1-2-4-18-15(3-1)20-19(25-18)22-9-7-21(8-10-22)12-14-5-6-16-17(11-14)24-13-23-16/h1-6,11H,7-10,12-13H2
SMIL C(N1CCN(CC1)c2nc3ccccc3s2)c4ccc5OCOc5c4
IUPAC navn 2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]-1,3-benzothiazole
Molekylvægt (g/mol) 353.438
Formel vægt 353.12