missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number 15455215
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1449135-43-6
C17H21BN2O5S
376.234
MFCD16294525
ACWASRSLONUFLW-UHFFFAOYSA-N
1,3-dimethyl-5-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethylene-pyrimidine-2,4,6-trione, 1,3-dimethyl-5-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methylidene-1,3-diazinane-2,4,6-trione, amtb203, 1,3-dimethyl-5-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-thienylmethylene pyrimidine-2,4,6-trione
71306514
1,3-dimethyl-5-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methylidene]-1,3-diazinane-2,4,6-trione
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C=C3C(=O)N(C(=O)N(C3=O)C)C
This item is not returnable. View return policy
1449135-43-6
C17H21BN2O5S
376.234
MFCD16294525
ACWASRSLONUFLW-UHFFFAOYSA-N
1,3-dimethyl-5-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethylene-pyrimidine-2,4,6-trione, 1,3-dimethyl-5-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methylidene-1,3-diazinane-2,4,6-trione, amtb203, 1,3-dimethyl-5-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-thienylmethylene pyrimidine-2,4,6-trione
71306514
1,3-dimethyl-5-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methylidene]-1,3-diazinane-2,4,6-trione
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C=C3C(=O)N(C(=O)N(C3=O)C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 376.23
Percent Purity 95%
Chemical Name or Material 2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol este
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'