Learn More
2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Spezifikation
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 164.12 |
Percent Purity | 97% |
Chemical Name or Material | 2, 1, 3-Benzoxadiazole-5-carboxylic acid |