Få mere at vide
2-(1,3-Benzothiazol-2-ylthio)succinic Acid, TRC
Artikelnummer. 30136881
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136881 100 mg
30136027 500 mg
30005446 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136881 Leverandør Toronto Research Chemicals Leverandørnr. TRCB130343100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 95154-01-1
Molekylær formel C11 H9 N O4 S2
Molekylvægt (g/mol) 283.32
Synonym (2-Benzothiazolylthio)succinic Acid, Butanedioic acid, 2-(2-benzothiazolylthio)-, Butanedioic acid, (2-benzothiazolylthio)- (9CI), 2-(2-Benzothiazolylthio)butanedioic acid, (2-Benzothiazolylthio)butanedioic acid, (2-Benzothiazolylthio)succinic acid, 2-(Benzo[d]thiazol-2-ylthio)succinic acid, 2-(Benzo[d]thiazol-2-ylthio)succinicacid, 2-(Benzothiazol-2-ylsulfanyl)succinic acid, Benzthiazol-2-ylthio succinic acid, Halox 650, Irgacor 252, Irgacor 252LD, Reocor 152
IUPAC navn 2-(1,3-benzothiazol-2-ylsulfanyl)butanedioic acid
SMIL OC(=O)CC(Sc1nc2ccccc2s1)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-(1,3-Benzothiazol-2-ylthio)succinic Acid
Mængde 100 mg
InChI formel InChI=1S/C11H9NO4S2/c13-9(14)5-8(10(15)16)18-11-12-6-3-1-2-4-7(6)17-11/h1-4,8H,5H2,(H,13,14)(H,15,16)
Formel vægt 282.9973