missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-carboxylic acid, 97%, Maybridge™

Brand:  Maybridge CC09101DE

Additional Details : CAS Number : 3529-57-5 Weight : 0.05500kg

Packaging Amber Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10462434

  • 5830.20 DKK / 5g

Estimated Shipment
28-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

3529-57-5
180.181
ZGDGZMOKXTUMEV-UHFFFAOYSA-N
2795202
C1=CC2=NSN=C2C(=C1)C(=O)O
C7H4N2O2S
MFCD05664666
benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
2,1,3-benzothiadiazole-4-carboxylic acid

Specifications

2, 1, 3-Benzothiadiazole-4-carboxylic acid
5g
96.5
97%
MFCD05664666
ZGDGZMOKXTUMEV-UHFFFAOYSA-N
2,1,3-benzothiadiazole-4-carboxylic acid
2795202
97%
Amber Glass Bottle
3529-57-5
100.0
C7H4N2O2S
benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
C1=CC2=NSN=C2C(=C1)C(=O)O
180.181
180.19
Videos
SDS Documents