missing translation for 'onlineSavingsMsg'
Få mere at vide

2-[1-(2-Naphthyl)ethyl]benzoic Acid, TRC

Artikelnummer. 30465615
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465615 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30465615 Leverandør Toronto Research Chemicals Leverandørnr. TRCN372050500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-[1-(2-Naphthyl)ethyl]benzoic Acid
CAS 904236-46-0
Molekylær formel C19 H16 O2
Mængde 500 mg
InChI formel InChI=1S/C19H16O2/c1-13(17-8-4-5-9-18(17)19(20)21)15-11-10-14-6-2-3-7-16(14)12-15/h2-13H,1H3,(H,20,21)
SMIL CC(c1ccc2ccccc2c1)c3ccccc3C(=O)O
IUPAC navn 2-(1-naphthalen-2-ylethyl)benzoic acid
Molekylvægt (g/mol) 276.33
Formel vægt 276.115
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.