Få mere at vide
(1S)-trans-Lambda-Cyhalothrin, TRC
Artikelnummer. 30469601
Skift visning
Mængde:
2.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469601 5 mg
30476954 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469601 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1700055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76703-69-0
Molekylær formel C23H19ClF3NO3
Molekylvægt (g/mol) 449.85
Synonym [1S-[1α(S*),3β(Z)]]-3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid Cyano(3-phenoxyphenyl)methyl Ester
IUPAC navn (R)-cyano(3-phenoxyphenyl)methyl (1S,3R)-3-((Z)-2-chloro-3,3,3-trifluoroprop-1-en-1-yl)-2,2-dimethylcyclopropane-1-carboxylate
SMIL O=C([C@@H]1C(C)(C)[C@H]1/C=C(Cl)/C(F)(F)F)O[C@@H](C#N)C2=CC(OC3=CC=CC=C3)=CC=C2
Tekniske data
Kemisk navn eller materiale (1S)-trans-Lambda-Cyhalothrin
Mængde 5 mg
InChI formel InChI=1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-/t17-,18?,20+/m0/s1
Formel vægt 449.1