missing translation for 'onlineSavingsMsg'
Få mere at vide

(1S)-(-)-Camphor, 98%

Artikelnummer. 11435467
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11435467 100 g 100g
11415467 5 g 5g
11425467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11435467 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B23469.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 464-48-2 | C10H16O | 152.24 g/mol

(1S)-(-)-Camphor is used as chiral intermediate and chiral auxiliary precursor. It is used in the synthesis of high-potency sweeteners. It acts as catalytic agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(-)-Camphor is used as chiral intermediate and chiral auxiliary precursor. It is used in the synthesis of high-potency sweeteners. It acts as catalytic agent and petrochemical additive.

Solubility
Soluble in water. (0.34 g/L) at 20°C

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 464-48-2
Molekylær formel C10H16O
Molekylvægt (g/mol) 152.24
MDL nummer MFCD00064148,MFCD00074738,MFCD00064149,MFCD00064149
InChI nøgle DSSYKIVIOFKYAU-OMNKOJBGSA-N
Synonym --camphor, l-camphor, 1s---camphor, 1s,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, --bornan-2-one, s-camphor, l--camphor, 1s,4s-camphor, camphor,-, --alcanfor
PubChem CID 444294
ChEBI CHEBI:15397
IUPAC navn (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMIL CC1(C)C2CC[C@]1(C)C(=O)C2

Tekniske data

Kemisk navn eller materiale (1S)-(-)-Camphor
Smeltepunkt 175°C to 180°C
Tæthed 0.99
Kogepunkt 204°C
Flammepunkt 65°C (149°F)
Lugt Camphor-like
Mængde 100 g
FN nummer UN2717
Beilstein 4291747
Merck Index 14,1732
Opløselighedsinformation Soluble in water. (0.34g/L) at 20°C
Optisk rotation −43° (c=10 in Ethanol)
Formel vægt 152.24
Procent renhed 98%
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.