Learn More
(1S)-(-)-Borneol, Sigma-Aldrich™
Product Code. 15839940
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839940 250 g 250g
15819940 25 g 25g
15829940 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15839940 Supplier Sigma Aldrich Supplier No. 8144720250
 Restricted Item
Forventet forsendelse: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 464-45-9
Grade Research
Molecular Weight (g/mol) 154.25
Specifications
Melting Point 205°C to 208°C
MDL Number MFCD00003759
Quantity 250 g
Synonym (1S)-(-)-Borneol, L-Borneol, (1S)-(-)-Borneol