missing translation for 'onlineSavingsMsg'
Få mere at vide

(1S)-(-)-(7,7-Dichloro-10-camphorsulfonyl)imine, TRC

Artikelnummer. 30387034
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30387034 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387034 Supplier Toronto Research Chemicals Supplier No. TRCD4339852.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (1S)-(-)-(7,7-Dichloro-10-camphorsulfonyl)imine
CAS 127184-04-7
Molekylær formel C10H13Cl2NO2S
Mængde 2.5 g
Synonym (3aS,6S)-7,7-Dichloro-4,5,6,7-tetrahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole 2,2-Dioxide
InChI formel InChI=1S/C10H13Cl2NO2S/c1-8(2)6-3-4-9(8)5-16(14,15)13-7(9)10(6,11)12/h6H,3-5H2,1-2H3
SMIL CC1(C)C2CCC13CS(=O)(=O)N=C3C2(Cl)Cl
Molekylvægt (g/mol) 282.19
Formel vægt 281.0044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.