Få mere at vide
(1S,4R)-1-Methyl-4-(1-methylethenyl)-2-cyclohexen-1-ol, TRC
Artikelnummer. 30476245
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476245 500 mg
30472612 1 g
30469706 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476245 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320460500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22972-51-6
Molekylær formel C10 H16 O
Molekylvægt (g/mol) 152.23
Synonym (1S-cis)-1-Methyl-4-(1-methylethenyl)-2-cyclohexen-1-ol, p-Mentha-2,8-dien-1-ol, (+)-(1S,4R)-p-Mentha-2,8-dien-1-ol, (+)-cis-p-Mentha-2,8-dien-1-ol, (+)-p-Mentha-2,8-dien-1-ol, (1S,4R)-p-Menth-2,8-dien-1-ol, cis-Isolimonenol, p-Mentha-2,8-dien-1-β-ol, PMD
IUPAC navn (1S,4R)-1-methyl-4-prop-1-en-2-ylcyclohex-2-en-1-ol
SMIL CC(=C)[C@@H]1CC[C@](C)(O)C=C1
Tekniske data
Kemisk navn eller materiale (1S,4R)-1-Methyl-4-(1-methylethenyl)-2-cyclohexen-1-ol
Mængde 500 mg
InChI formel InChI=1S/C10H16O/c1-8(2)9-4-6-10(3,11)7-5-9/h4,6,9,11H,1,5,7H2,2-3H3/t9-,10+/m0/s1
Formel vægt 152.1201