Få mere at vide
1S,3R-RSL3, Tocris Bioscience™
GREENER_CHOICE
Inhibits glutathione peroxidase (GPX4); induces ferroptosis
Brand:  Tocris Bioscience 6118/10
Encompass
Produktkode 15923261
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.
Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.
Tekniske data
Tekniske data
(1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate
1219810-16-8
>98%
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner