Få mere at vide
1S,3R-RSL3, Tocris Bioscience™
Inhibits glutathione peroxidase (GPX4); induces ferroptosis
Brand: Tocris Bioscience 6118/50
Description
1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.
Specifications
| (1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate | |
| 1219810-16-8 | |
| >98% | |
| 50 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.