Få mere at vide
(1S,3R,4R)-3-[(tert-Butoxycarbonyl)amino]-4-[(methylsulfonyl)oxy]cyclohexanecarboxylic Acid Ethyl Ester, TRC
Artikelnummer. 30347646
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347646 5 g
30334721 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30347646 Leverandør Toronto Research Chemicals Leverandørnr. TRCB7500555G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1S,3R,4R)-3-[(tert-Butoxycarbonyl)amino]-4-[(methylsulfonyl)oxy]cyclohexanecarboxylic Acid Ethyl Ester
CAS 365997-36-0
Molekylær formel C15H27NO7S
Mængde 5 g
Synonym (1S,3R,4R)-3-[(tert-Butoxycarbonyl)amino]-4-[(methanesulfonyl)oxy]cyclohexanecarboxylic Acid Ethyl Ester
InChI formel InChI=1S/C15H27NO7S/c1-6-21-13(17)10-7-8-12(23-24(5,19)20)11(9-10)16-14(18)22-15(2,3)4/h10-12H,6-9H2,1-5H3,(H,16,18)/t10-,11+,12+/m0/s1
SMIL CCOC(=O)[C@H]1CC[C@@H](OS(=O)(=O)C)[C@@H](C1)NC(=O)OC(C)(C)C
IUPAC navn ethyl (1S,3R,4R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfonyloxycyclohexane-1-carboxylate
Molekylvægt (g/mol) 365.44
Formel vægt 365.1508