missing translation for 'onlineSavingsMsg'
Få mere at vide

(1S,3R,4R)-3-[(tert-Butoxycarbonyl)amino]-4-[(methylsulfonyl)oxy]cyclohexanecarboxylic Acid Ethyl Ester, TRC

Artikelnummer. 30334721
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334721 1 g
30347646 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30334721 Leverandør Toronto Research Chemicals Leverandørnr. TRCB7500551G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (1S,3R,4R)-3-[(tert-Butoxycarbonyl)amino]-4-[(methylsulfonyl)oxy]cyclohexanecarboxylic Acid Ethyl Ester
CAS 365997-36-0
Molekylær formel C15H27NO7S
Mængde 1 g
Synonym (1S,3R,4R)-3-[(tert-Butoxycarbonyl)amino]-4-[(methanesulfonyl)oxy]cyclohexanecarboxylic Acid Ethyl Ester
InChI formel InChI=1S/C15H27NO7S/c1-6-21-13(17)10-7-8-12(23-24(5,19)20)11(9-10)16-14(18)22-15(2,3)4/h10-12H,6-9H2,1-5H3,(H,16,18)/t10-,11+,12+/m0/s1
SMIL CCOC(=O)[C@H]1CC[C@@H](OS(=O)(=O)C)[C@@H](C1)NC(=O)OC(C)(C)C
IUPAC navn ethyl (1S,3R,4R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfonyloxycyclohexane-1-carboxylate
Molekylvægt (g/mol) 365.44
Formel vægt 365.1508
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.