Få mere at vide
(1S,2S)-N-Acetylpseudoephedrine, Mikromol™
Artikelnummer. 30050548
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050548 25 mg
30052115 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050548 Leverandør Mikromol Leverandørnr. MM2115.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1S,2S)-N-Acetylpseudoephedrine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1S,2S)-N-Acetylpseudoephedrine
CAS 84472-25-3
Analyt- eller komponentnavne (1S,2S)-N-Acetylpseudoephedrine
Forsendelsestilstand Room Temperature
Molekylær formel C12 H17 N O2
Certificeringer/compliance ISO 17025
Synonym (1S,2S)-N-Acetylpseudoephedrine
InChI formel InChI=1S/C12H17NO2/c1-9(13(3)10(2)14)12(15)11-7-5-4-6-8-11/h4-9,12,15H,1-3H3/t9-,12+/m0/s1
SMIL C[C@@H]([C@@H](O)c1ccccc1)N(C)C(=O)C
IUPAC navn N-[(1S,2S)-1-hydroxy-1-phenylpropan-2-yl]-N-methylacetamide
Molekylvægt (g/mol) 207.27
Mængde 25 mg
Formel vægt 207.1259 g/mol