missing translation for 'onlineSavingsMsg'
Få mere at vide

(1S,2S)-Bortezomib, TRC

Artikelnummer. 30337384
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337384 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30337384 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6757151MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (1S,2S)-Bortezomib
CAS 1132709-14-8
Molekylær formel C19 H25 B N4 O4
Mængde 1 mg
Synonym B-[(1S)-3-Methyl-1-[[(2S)-1-oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]butyl]-boronic Acid, bortezomib diastereomer (S,S) (USP)
InChI formel InChI=1S/C19H25BN4O4/c1-13(2)10-17(20(27)28)24-18(25)15(11-14-6-4-3-5-7-14)23-19(26)16-12-21-8-9-22-16/h3-9,12-13,15,17,27-28H,10-11H2,1-2H3,(H,23,26)(H,24,25)/t15-,17+/m0/s1
SMIL O=C(NC(C(=O)NC(B(O)O)CC(C)C)CC=1C=CC=CC1)C2=NC=CN=C2
IUPAC navn [(1S)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
Molekylvægt (g/mol) 384.24
Formel vægt 384.1969
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.