Få mere at vide
(1S,2S,3R,5S)-3-[[3-[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]-5-(propylthio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-7-yl]amino]-5-(2-hydroxyethoxy)-1,2-cyclopentanediol, TRC
Artikelnummer. 30380056
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30380056 10 mg
30377117 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30380056 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44533510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1788033-05-5
Molekylær formel C23H28F2N6O4S
Molekylvægt (g/mol) 522.57
Synonym Ticagrelor Imp A
SMIL CCCSc1nc(N[C@@H]2C[C@H](OCCO)[C@@H](O)[C@H]2O)c3nnn([C@@H]4C[C@H]4c5ccc(F)c(F)c5)c3n1
Tekniske data
Kemisk navn eller materiale (1S,2S,3R,5S)-3-((3-((1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl)-5-(propylthio)-3H-1,2,3-triazolo(4,5-d)pyrimidin-7-yl)amino)-5-(2-hydroxyethoxy)-1,2-cyclopentanediol
Mængde 10 mg
InChI formel InChI=1S/C23H28F2N6O4S/c1-2-7-36-23-27-21(26-15-10-17(35-6-5-32)20(34)19(15)33)18-22(28-23)31(30-29-18)16-9-12(16)11-3-4-13(24)14(25)8-11/h3-4,8,12,15-17,19-20,32-34H,2,5-7,9-10H2,1H3,(H,26,27,28)/t12-,15+,16+,17-,19-,20+/m0/s1
Formel vægt 522.1861