Få mere at vide
(1S,2S)-1,2-Cyclopropanedicarboxylic Acid 1,2-Diethyl Ester, TRC
Artikelnummer. 30359146
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359146 500 mg
30351887 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359146 Leverandør Toronto Research Chemicals Leverandørnr. TRCC990413500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1S,2S)-1,2-Cyclopropanedicarboxylic Acid 1,2-Diethyl Ester
CAS 889461-57-8
Molekylær formel C9 H14 O4
Mængde 500 mg
Synonym (1S,2S)-Diethyl cyclopropane-1,2-dicarboxylate
InChI formel InChI=1S/C9H14O4/c1-3-12-8(10)6-5-7(6)9(11)13-4-2/h6-7H,3-5H2,1-2H3/t6-,7-/m0/s1
SMIL CCOC(=O)[C@H]1C[C@@H]1C(=O)OCC
IUPAC navn diethyl (1S,2S)-cyclopropane-1,2-dicarboxylate
Molekylvægt (g/mol) 186.205
Formel vægt 186.089