Få mere at vide
(1S,2R)-Ticagrelor, TRC
Artikelnummer. 30379547
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379547 25 mg
30377904 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30379547 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44589125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2096989-56-7
Molekylær formel C23H28F2N6O4S
Molekylvægt (g/mol) 522.57
Synonym Ticagrelor Impurity, (1S,2R)-Ticagrelor
SMIL CCCSc1nc(N[C@H]2C[C@@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCO)[C@@H](O)[C@H]5O)c4n1
Tekniske data
Kemisk navn eller materiale (1S,2S,3R,5S)-3-(7-(((1S,2R)-2-(3,4-Difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-5-(2-hydroxyethoxy)cyclopentane-1,2-diol
Mængde 25 mg
InChI formel InChI=1S/C23H28F2N6O4S/c1-2-7-36-23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(35-6-5-32)20(34)19(16)33/h3-4,8,12,15-17,19-20,32-34H,2,5-7,9-10H2,1H3,(H,26,27,28)/t12-,15+,16-,17+,19+,20-/m1/s1
Formel vægt 522.1861