missing translation for 'onlineSavingsMsg'
Learn More
(1S,2R)-(-)-cis-1-Amino-2-indanol, 97% Product Code.: 15424747

(1S,2R)-(-)-cis-1-Amino-2-indanol, 97%

Product Code. 15424747
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15424747

Brand: Thermo Scientific Alfa Aesar H32066.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126456-43-7 | C9H12NO | 150.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126456-43-7
Molecular Formula C9H12NO
Molecular Weight (g/mol) 150.20
MDL Number MFCD00216655
InChI Key LOPKSXMQWBYUOI-BDAKNGLRSA-O
Synonym 1s,2r-1-amino-2,3-dihydro-1h-inden-2-ol, 1s,2r---cis-1-amino-2-indanol, 1s,2r---1-amino-2-indanol, 1s,2r-1-amino-2-indanol, 1s,2r-cis-1-amino-2-indanol, cis-1s,2r-1-amino-2-indanol, 1s,2r-1-amino-indan-2-ol, 1s,2r---cis-1-aminoindan-2-ol, 1s,2r---1-amino-2-hydroxyindan, 1s, 2r-cis-1-amino-2-indanol
PubChem CID 9866743
IUPAC Name (1S,2R)-1-amino-2,3-dihydro-1H-inden-2-ol
SMILES [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12

Specifications

Melting Point 117°C to 118°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Optical Rotation −61° (c=0.5 in Chloroform)
Formula Weight 149.19
Percent Purity 97%
Chemical Name or Material (1S,2R)-(-)-cis-1-Amino-2-indanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1S,2R)-(-)-cis-1-Amino-2-indanol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.