Få mere at vide
[(1S,2R)-1-Benzyl-2-hydroxy-3-[isobutyl-d9-[(4-nitrophenyl)sulfonyl]amino]propyl]carbamic Acid tert-Butyl Ester, TRC
Artikelnummer. 30345638
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30345638 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345638 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2790721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale [(1S,2R)-1-Benzyl-2-hydroxy-3-[isobutyl-d9-[(4-nitrophenyl)sulfonyl]amino]propyl]carbamic Acid tert-Butyl Ester
CAS 1146967-62-5
Molekylær formel C25H26D9N3O7S
Mængde 1 mg
Synonym N-[(1S,2R)-2-Hydroxy-3-[(2-methylpropyl-d9)[(4-nitrophenyl)sulfonyl]amino]-1-(phenylmethyl)propyl]carbamic Acid 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C25H35N3O7S/c1-18(2)16-27(36(33,34)21-13-11-20(12-14-21)28(31)32)17-23(29)22(15-19-9-7-6-8-10-19)26-24(30)35-25(3,4)5/h6-14,18,22-23,29H,15-17H2,1-5H3,(H,26,30)/t22-,23+/m0/s1/i1D3,2D3,16D2,18D
SMIL [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])N(C[C@@H](O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C)S(=O)(=O)c2ccc(cc2)[N+](=O)[O-]
IUPAC navn tert-butyl N-[(2S,3R)-4-[[1,1,2,3,3,3-hexadeuterio-2-(trideuteriomethyl)propyl]-(4-nitrophenyl)sulfonylamino]-3-hydroxy-1-phenylbutan-2-yl]carbamate
Molekylvægt (g/mol) 530.68