Få mere at vide
(1RS,2SR)-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol Hydrochloride, Mikromol™
Beskrivelse
(1RS,2SR)-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (1RS,2SR)-2-((Dimethylamino)methyl)-1-(3-methoxyphenyl)cyclohexanol Hydrochloride |
| CAS | 73806-49-2 |
| Analyt- eller komponentnavne | (1RS,2SR)-2-((Dimethylamino)methyl)-1-(3-methoxyphenyl)cyclohexanol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H25 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Tramadol Hydrochloride Imp. A (EP), Tramadol Imp. A (EP), Tramadol USP Related Compound A CIV, Tramadol USP RC A CIV, (1RS,2SR)-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol Hydrochloride, Tramadol Hydrochloride Imp. A (EP) as Hydrochloride, Tramadol Related Compound A as Hydrochloride, Tramadol Hydrochloride Impurity A as Hydrochloride, Tramadol Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16-;/m0./s1 |
| SMIL | Cl.COc1cccc(c1)[C@@]2(O)CCCC[C@H]2CN(C)C |
| IUPAC navn | (1R,2S)-2-[(dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexan-1-ol;hydrochloride |
| Vis mere |