Få mere at vide
(1RS)-[2-(3-Methoxyphenyl)cyclohex-2-enyl]-N,N-dimethylmethanamine Hydrochloride, Mikromol™
Beskrivelse
(1RS)-[2-(3-Methoxyphenyl)cyclohex-2-enyl]-N,N-dimethylmethanamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Anhydrotramadol Hydrochloride |
| CAS | 66170-31-8 |
| Analyt- eller komponentnavne | 2-Anhydrotramadol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H23 N O . H Cl |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Tramadol Hydrochloride Imp. C (EP), Tramadol Imp. C (EP), (1RS)-[2-(3-Methoxyphenyl)cyclohex-2-enyl]-N,N-dimethylmethanamine Hydrochloride, Tramadol Hydrochloride Imp. C (EP) as Hydrochloride, 2-Anhydrotramadol Hydrochloride, 2-(3-Methoxyphenyl)-N,N-dimethyl-2-cyclohexene-1-methanamine Hydrochloride, Tramadol Hydrochloride Impurity C as Hydrochloride, Tramadol Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C16H23NO.ClH/c1-17(2)12-14-7-4-5-10-16(14)13-8-6-9-15(11-13)18-3;/h6,8-11,14H,4-5,7,12H2,1-3H3;1H |
| SMIL | Cl.COc1cccc(c1)C2=CCCCC2CN(C)C |
| IUPAC navn | 1-[2-(3-methoxyphenyl)cyclohex-2-en-1-yl]-N,N-dimethyl-methanamine;hydrochloride |
| Vis mere |