Få mere at vide
(1R)-trans-Gamma-Cyhalothrin, TRC
Artikelnummer. 30367339
Skift visning
Mængde:
2.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367339 5 mg
30349077 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367339 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9898505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76703-66-7
Molekylær formel C23H19ClF3NO3
Molekylvægt (g/mol) 449.85
Synonym (1R,3S)-3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-cyclopropanecarboxylic Acid (S)-cyano(3-phenoxyphenyl)methyl Ester, [1R-[1α(S*),3β(Z)]]-3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-cyclopropanecarboxylic Acid (S)-cyano(3-phenoxyphenyl)methyl Ester, (-)-(1R)-trans-γ-Cyhalothrin
IUPAC navn (S)-cyano(3-phenoxyphenyl)methyl (1R,3S)-3-((Z)-2-chloro-3,3,3-trifluoroprop-1-en-1-yl)-2,2-dimethylcyclopropane-1-carboxylate
SMIL FC(F)(F)/C(Cl)=C/[C@@H]1[C@H](C1(C)C)C(O[C@@H](C2=CC(OC3=CC=CC=C3)=CC=C2)C#N)=O
Tekniske data
Kemisk navn eller materiale (1R)-trans-gamma-Cyhalothrin
Mængde 5 mg
InChI formel InChI=1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-/t17-,18-,20+/m1/s1
Formel vægt 449.1