Få mere at vide
(1R,4S,4aR,9aS)-rel-1,4,4a,9a-Tetrahydro-4a-methyl-1,4-methanoanthracene-9,10-dione-d8, TRC
Artikelnummer. 30414395
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30414395 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414395 Leverandør Toronto Research Chemicals Leverandørnr. TRCT293982100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1R,4S,4aR,9aS)-rel-1,4,4a,9a-Tetrahydro-4a-methyl-1,4-methanoanthracene-9,10-dione-d8
CAS 1329799-73-6
Molekylær formel C16H6D8O2
Mængde 100 mg
Synonym (1alpha,4alpha,4aalpha,9aalpha)-1,4,4a,9a-Tetrahydro-4a-methyl-1,4-methanoanthracene-9,10-dione-d8
InChI formel InChI=1S/C16H14O2/c1-16-10-7-6-9(8-10)13(16)14(17)11-4-2-3-5-12(11)15(16)18/h2-7,9-10,13H,8H2,1H3/t9-,10+,13+,16+/m0/s1/i1D3,2D,3D,4D,5D,13D
SMIL [2H]c1c([2H])c([2H])c2C(=O)[C@@]3([C@H]4C[C@H](C=C4)[C@]3([2H])C(=O)c2c1[2H])C([2H])([2H])[2H]
Molekylvægt (g/mol) 246.33
Formel vægt 246.1496