missing translation for 'onlineSavingsMsg'
Få mere at vide

(1R,3S,5R)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid, Benzyl Ester, TRC

Artikelnummer. 30329229
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30329229 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30329229 Leverandør Toronto Research Chemicals Leverandørnr. TRCA79527550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale benzyl (2S,3aR,6aR)-1,2,3,3a,4,5,6,6a-octahydrocyclopenta[b]pyrrole-2-carboxylate
CAS 130609-48-2
Molekylær formel C15 H19 N O2
Mængde 50 mg
InChI formel InChI=1S/C15H19NO2/c17-15(18-10-11-5-2-1-3-6-11)14-9-12-7-4-8-13(12)16-14/h1-3,5-6,12-14,16H,4,7-10H2/t12-,13-,14+/m1/s1
SMIL O=C(OCc1ccccc1)[C@@H]2C[C@H]3CCC[C@H]3N2
IUPAC navn benzyl (2S,3aR,6aR)-1,2,3,3a,4,5,6,6a-octahydrocyclopenta[b]pyrrole-2-carboxylate
Molekylvægt (g/mol) 245.32
Formel vægt 245.1416
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.