Få mere at vide
(1R,3S)-3-Aminocyclopentanol Hydrochloride, TRC
Artikelnummer. 30327365
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327365 50 mg
30327082 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30327365 Leverandør Toronto Research Chemicals Leverandørnr. TRCA59654050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1R,3S)-3-Aminocyclopentanol Hydrochloride
CAS 1279032-31-3
Molekylær formel C5 H11 N O . Cl H
Mængde 50 mg
Synonym (1R,3S)-3-Aminocyclopentan-1-ol Hydrochloride
InChI formel InChI=1S/C5H11NO.ClH/c6-4-1-2-5(7)3-4;/h4-5,7H,1-3,6H2;1H/t4-,5+;/m0./s1
SMIL Cl.N[C@H]1CC[C@@H](O)C1
IUPAC navn (1R,3S)-3-aminocyclopentanol;hydrochloride
Molekylvægt (g/mol) 137.61
Formel vægt 137.0607