Få mere at vide
(1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate, TRC
Artikelnummer. 30473727
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473727 5 g
30475850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473727 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2204005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate
CAS 1517-82-4
Molekylær formel C17H26O2S
Mængde 5 g
Synonym [1R-[1α(S*),2β,5α]]-4-Methylbenzenesulfinic Acid 5-Methyl-2-(1-methylethyl)cyclohexyl Ester, (1R,3R,4S)-Menthol (-)-(S)-p-Toluenesulfinate, (S)-p-Toluenesulfinic Acid (-)-(1R,3R,4S)-p-Menth-3-yl Ester, (-)-(1R)-Menthyl (S)-toluene-4-sulfinate
InChI formel InChI=1S/C17H26O2S/c1-12(2)16-10-7-14(4)11-17(16)19-20(18)15-8-5-13(3)6-9-15/h5-6,8-9,12,14,16-17H,7,10-11H2,1-4H3/t14-,16+,17-,20+/m1/s1
SMIL C[C@H]1C[C@@H](O[S@@](C2=CC=C(C)C=C2)=O)[C@H](C(C)C)CC1
IUPAC navn (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl (S)-4-methylbenzenesulfinate
Molekylvægt (g/mol) 294.45
Formel vægt 294.17