Få mere at vide
(1R,2S,5R)-(-)-Menthol-d4, TRC
Artikelnummer. 30470562
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30470562 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470562 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2188771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1R,2S,5R)-(-)-Menthol-d4
CAS 1450828-98-4
Molekylær formel C10H16D4O
Mængde 1 mg
Synonym (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol-d4, [1R-(1α,2β,5α)]-5-Methyl-2-
InChI formel InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1/i6D2,9D,10D
SMIL O[C@@]1([2H])[C@@](C(C)C)([2H])CC[C@@H](C)C1([2H])[2H]
IUPAC navn (1R,2S,5R)-2-isopropyl-5-methylcyclohexan-1,2,6,6-d4-1-ol
Molekylvægt (g/mol) 160.29
Formel vægt 160.18