Få mere at vide
(1R,2R,4S,5S,7s)-7-[[(2S)-3-Hydroxy-2-phenylpropanoyl]oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide (Methylhyoscine Bromide), Mikromol™
Beskrivelse
(1R,2R,4S,5S,7s)-7-[[(2S)-3-Hydroxy-2-phenylpropanoyl]oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide (Methylhyoscine Bromide)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methscopolamine Bromide |
| CAS | 155-41-9 |
| Analyt- eller komponentnavne | Methscopolamine Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H24 N O4 . Br |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Homatropine Methylbromide Imp. D (EP), (1R,2R,4S,5S,7s)-7-[[(2S)-3-Hydroxy-2-phenylpropanoyl]oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide, Homatropine Methylbromide Imp. D (EP) as Bromide, Methylhyoscine Bromide, Oxitropium bromide EP impurity B, Homatropine Methylbromide Impurity D as Bromide |
| InChI formel | InChI=1S/C18H24NO4.BrH/c1-19(2)14-8-12(9-15(19)17-16(14)23-17)22-18(21)13(10-20)11-6-4-3-5-7-11;/h3-7,12-17,20H,8-10H2,1-2H3;1H/q+1;/p-1/t12-,13-,14-,15+,16-,17+;/m1./s1 |
| SMIL | [Br-].C[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4 |
| IUPAC navn | (1R,2R,4S,5S,7s)-7-[[(2S)-3-Hydroxy-2-phenylpropanoyl]oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide |
| Vis mere |