missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%, Thermo Scientific Chemicals

Catalog Number p-4229258
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
69460-11-3
C10H19N
153.27
VPTSZLVPZCTAHZ-BZNPZCIMSA-N
1r,2r,3r,5s---isopinocampheylamine, 1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine, bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s, --isopinocampheylamine, --isopinocampheyl amine, 1r,2r,3r,5s-3-pinanamine, 1r,2r,5s-pinane-3alpha-amine, --isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine, 1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine
11744832
(1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
CC1C2CC(C2(C)C)CC1N
This item is not returnable. View return policy
69460-11-3
C10H19N
153.27
VPTSZLVPZCTAHZ-BZNPZCIMSA-N
1r,2r,3r,5s---isopinocampheylamine, 1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine, bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s, --isopinocampheylamine, --isopinocampheyl amine, 1r,2r,3r,5s-3-pinanamine, 1r,2r,5s-pinane-3alpha-amine, --isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine, 1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine
11744832
(1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
CC1C2CC(C2(C)C)CC1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 90°C (18.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -42
Formula Weight 153.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (1R,2R,3R,5S)-(-)-Isopinocampheylamine
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2R,3R,5S)-(-)-Isopinocampheylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'