Få mere at vide
(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride |
| CAS | 158414-44-9 |
| Molekylær formel | C6H11NO2 . HCl |
| Mængde | 10 mg |
| Synonym | Cyclopentanecarboxylic acid, 2-amino-, hydrochloride, (1R-trans)- (9CI), (1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride, (1R,2R)-2-aminocyclopentanecarboxylic acid hydrochloride (1:1), (1R,2R)-2-Aminocyclopentane-1-carboxylic acid hydrochloride, trans-(1R,2R)-2-Aminocyclopentane-1-carboxylic acid hydrochloride, trans-(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride |
| InChI formel | InChI=1S/C6H11NO2.ClH/c7-5-3-1-2-4(5)6(8)9;/h4-5H,1-3,7H2,(H,8,9);1H/t4-,5-;/m1./s1 |
| SMIL | Cl.N[C@@H]1CCC[C@H]1C(=O)O |
| IUPAC navn | (1R,2R)-2-aminocyclopentane-1-carboxylic acid;hydrochloride |
| Molekylvægt (g/mol) | 165.62 |
| Formel vægt | 165.0557 |