Learn More
1LT CHLOROFORM BAKER ANALYZED Reagent
Brand:  JT Baker 7071.1000
1391.75 DKK valid until 2024-12-31
Use promo code "23268" to get your promotional price.
1LT CHLOROFORM BAKER ANALYZED Reagent
Additional Details : CAS Number : 67-66-3 Weight : 1.00000kg Transport : UN number : 1888 Chem class : 6.1 Pack group : III
Product Code. 13498509
  • 1465.00 DKK / 1 L
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P281-Use personal protective equipment as required.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.